5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A709788-1G A709788-5G A709788-100MG A709788-250MG
77684-625EA 564 USD
77684-625 77891-394 77891-392 77891-393
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile ≥98%, Ambeed.Inc
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile

Precaución: For research use only.

Formula: C₁₄H₁₅BN₂O₂
MDL Number: MFCD08061937
CAS Number: 863868-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR