9-Methoxy-4-((3-methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one ≥98%

Supplier: Ambeed
Warning

Synonyms: CNIDILIN

A709673-10MG A709673-1MG
77598-740EA 322.88 USD
77598-740 77598-742
9-Methoxy-4-((3-methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one ≥98%
9-Methoxy-4-((3-methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
MDL Number: MFCD09752956
CAS Number: 14348-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR