3-Azabicyclo[3.2.1]octane hydrochloride ≥95%

Supplier: Ambeed
Warning

A709006-100MG A709006-1G
77617-926EA 177.16 USD
77617-926 77749-451
3-Azabicyclo[3.2.1]octane hydrochloride ≥95%
3-Azabicyclo[3.2.1]octane hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClN
MW: 147.65 g/mol
MDL Number: MFCD22196503
CAS Number: 20969-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR