1-Isocyanoadamantane ≥97% (by GC)

Supplier: Ambeed
Danger

Synonyms: 1-Adamantyl Isocyanide

A708505-1G A708505-250MG
77783-696EA 143 USD
77783-696 77783-697
1-Isocyanoadamantane ≥97% (by GC)
1-Isocyanoadamantane

Precaución: for Research Use Only

Formula: C₁₁H₁₅N
MDL Number: MFCD02258889
CAS Number: 22110-53-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR