3-Bromo-2-methylpyridine-5-boronic acid pinacol ester ≥96%

Supplier: Ambeed
Warning

A708367-5G A708367-1G A708367-250MG
77622-518EA 610.55 USD
77622-518 77783-694 77783-695
3-Bromo-2-methylpyridine-5-boronic acid pinacol ester ≥96%
3-Bromo-2-methylpyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BBrNO₂
MDL Number: MFCD16995186
CAS Number: 2223051-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR