2-Chloro-3-chloromethyl-pyridine ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-3-(chloromethyl)pyridine

A707506-10G A707506-1G A707506-5G
77673-934EA 156 USD
77673-934 77673-936 77673-938
2-Chloro-3-chloromethyl-pyridine ≥98%
2-Chloro-3-chloromethyl-pyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.017 g/mol
MDL Number: MFCD09835107
CAS Number: 89581-84-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR