Magnesium citrate ≥98% (contains 20-32% H₂O)

Supplier: Ambeed
Warning

Synonyms: Trimagnesium dicitrate, Magnesium 2-hydroxypropane-1,2,3-tricarboxylate

A707169-500G A707169-25G
77622-516EA 36.05 USD
77622-516 77783-693
Magnesium citrate ≥98% (contains 20-32% H₂O)
Magnesium citrate

Precaución: for Research Use Only

Formula: C₆H₈O₇·nMg
MDL Number: MFCD00036422
CAS Number: 7779-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR