Boc-O-benzyl-L-β-homoserine ≥98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-HomoSer(Bzl)-OH, Boc-β-HoSer(Bzl)-OH, Boc-β-Hse(Bzl)-OH

A707065-100MG A707065-250MG
77882-567EA 14 USD
77882-567 77882-568
Boc-O-benzyl-L-β-homoserine ≥98%
Boc-O-benzyl-L-beta-homoserine

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MW: 309.37 g/mol
MDL Number: MFCD01862866
CAS Number: 218943-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR