9-Amino-1-nonanol ≥95%

Supplier: Ambeed
Danger

Synonyms: 9-Aminononan-1-ol

A706998-1G A706998-250MG A706998-100MG
77598-730EA 261.38 USD
77598-730 77598-732 77783-692
9-Amino-1-nonanol ≥95%
9-Amino-1-nonanol

Precaución: for Research Use Only

Formula: C₉H₂₁NO
MDL Number: MFCD02094312
CAS Number: 109055-42-7
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR