Fmoc-O-trityl-D-serine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-O-trityl-D-serine, Fmoc-D-Ser(Trt)-OH
A706948-25G
A706948-5G
A706948-100G
77573-294EA
65
USD
77573-294
77673-932
77882-566
Fmoc-O-trityl-D-serine ≥98%
Fmoc-O-trityl-D-serine
Precaución: for Research Use Only
Formula:
C₃₇H₃₁NO₅ |
MDL Number:
MFCD00270546 CAS Number: 212688-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...