Nickel(II) sulfide 99.9%

Supplier: Ambeed
Danger

A706909-5G A706909-25G
77622-514EA 37.08 USD
77622-514 77783-691
Nickel(II) sulfide 99.9%
Nickel(II) sulfide

Precaución: for Research Use Only

Formula: NiS
MDL Number: MFCD00016266
CAS Number: 16812-54-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR