4-(Methylamino)pyrrolidin-2-one ≥98%

Supplier: Ambeed
Warning

A706767-250MG A706767-100MG
77573-292EA 565.47 USD
77573-292 77749-450
4-(Methylamino)pyrrolidin-2-one ≥98%
4-(Methylamino)pyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O
MW: 114.1457 g/mol
MDL Number: MFCD28132784
CAS Number: 1195687-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR