Methyl-5-amino-2-bromo-4-chlorobenzoate ≥98%

Supplier: Ambeed
Warning

A706751-1G A706751-5G
77884-619EA 25 USD
77884-619 77884-620
Methyl-5-amino-2-bromo-4-chlorobenzoate ≥98%
Methyl 5-amino-2-bromo-4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MDL Number: MFCD12546470
CAS Number: 929524-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR