6-Methoxypyridine-3-sulfonyl chloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 6-Methoxy-pyridine-3-sulphonyl chloride, 3-Pyridinesulphonylchloride, 6-methoxy-, 3-(Chlorosulphonyl)-6-methoxypyridine

A706478-1G A706478-250MG
77684-620EA 48 USD
77684-620 77891-386
6-Methoxypyridine-3-sulfonyl chloride ≥95%, Ambeed.Inc
6-Methoxypyridine-3-sulfonyl chloride

Precaución: For research use only.

Formula: C₆H₆ClNO₃S
MDL Number: MFCD06739108
CAS Number: 312300-42-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR