1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione mixt. with 5-chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione (1:1) hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Trifluridine/tipiracil HCl mixture

A705747-10MG A705747-1MG A705747-5MG
77617-910EA 41.2 USD
77617-910 77617-912 77617-914
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione mixt. with 5-chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione (1:1) hydrochloride ≥98%
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione mixt. with 5-chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione (1:1) hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₃Cl₂F₃N₆O₇
MW: 575.32 g/mol
MDL Number: MFCD30481309
CAS Number: 733030-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR