Adenosine 5'-monophosphate monohydrate ≥95%

Supplier: Ambeed
Warning

Synonyms: A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid

A705530-25G A705530-1G A705530-5G
77602-802EA 26.65 USD
77602-802 77627-440 77627-442
Adenosine 5'-monophosphate monohydrate ≥95%
Adenosine 5'-monophosphate monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅O₇P·1H₂O
MW: 365.24 g/mol
MDL Number: MFCD00149360
CAS Number: 18422-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR