4-((3S,5S,8R,9S,10R,13R,14S,17R)-5,14-Dihydroxy-3-(((2R,4S,5R,6R)-4-methoxy-6-methyl-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)furan-2(5H)-one ≥97%

Supplier: Ambeed
Warning

A705257-1MG A705257-5MG A705257-10MG
77669-896EA 9 USD
77669-896 77669-898 77880-463
4-((3S,5S,8R,9S,10R,13R,14S,17R)-5,14-Dihydroxy-3-(((2R,4S,5R,6R)-4-methoxy-6-methyl-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)furan-2(5H)-one ≥97%
4-((3S,5S,8R,9S,10R,13R,14S,17R)-5,14-Dihydroxy-3-(((2R,4S,5R,6R)-4-methoxy-6-methyl-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren

Precaución: for Research Use Only

Formula: C₃₆H₅₆O₁₃
MDL Number: MFCD01745141
CAS Number: 13137-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR