6-Fluoro-1H-pyrrolo[3,2-b]pyridin-5-amine ≥96%

Supplier: Ambeed
Warning

A704755-100MG A704755-1G
77749-444EA 320 USD
77749-444 77749-445
6-Fluoro-1H-pyrrolo[3,2-b]pyridin-5-amine ≥96%
6-Fluoro-1H-pyrrolo[3,2-b]pyridin-5-amine

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.14 g/mol
MDL Number: MFCD31705036
CAS Number: 2231676-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR