m-PEG8-NHS ester ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl 4,7,10,13,16,19,22,25-Octaoxahexacosanoate, 2,5-Dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23-octaoxahexacosan-26-oate, Methyl-PEG8-NHS Ester, mPEG8-NHS Ester, 4,7,10,13,16,19,22,25-Octaoxahexacosanoic Acid N-Succinimidyl Ester

A703316-1G A703316-250MG
77602-798EA 218.33 USD
77602-798 77686-689
m-PEG8-NHS ester ≥98%
m-PEG8-NHS ester

Precaución: for Research Use Only

Formula: C₂₂H₃₉NO₁₂
MW: 509.54 g/mol
MDL Number: MFCD13184969
CAS Number: 756525-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR