4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperidine, 4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester

A702670-1G A702670-250MG A702670-5G
77891-380EA 43 USD
77891-380 77891-381 77891-382
4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester ≥98%
4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₂
MW: 301.23 g/mol
MDL Number: MFCD08271932
CAS Number: 859833-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR