5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic acid methyl ester ≥98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Eicosapentaenoic acid methyl ester, (5Z,8Z,11Z,14Z,17Z)-Methyl icosa-5,8,11,14,17-pentaenoate

A701939-1G A701939-100MG A701939-5G
77602-786EA 126.08 USD
77602-786 77627-434 77627-436
5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic acid methyl ester ≥98% (by GC)
5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic acid methyl ester

Precaución: for Research Use Only

Formula: C₂₁H₃₂O₂
MW: 316.485 g/mol
MDL Number: MFCD02259226
CAS Number: 2734-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR