2,5,8,11,14-Pentaoxahexadecan-16-oic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: m-PEG4-CH2COOH

A701671-1G A701671-250MG A701671-5G
77598-724EA 129.15 USD
77598-724 77622-500 77622-502
2,5,8,11,14-Pentaoxahexadecan-16-oic acid ≥95%
2,5,8,11,14-Pentaoxahexadecan-16-oic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₇
MW: 266.2882 g/mol
MDL Number: MFCD00021759
CAS Number: 16024-66-1
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR