4-Nitrophenyl-N-acetyl-b-D-galactosaminide ≥98%

Supplier: Ambeed
Warning

A700641-100MG A700641-1G A700641-250MG
77617-892EA 99.59 USD
77617-892 77617-894 77617-896
4-Nitrophenyl-N-acetyl-b-D-galactosaminide ≥98%
4-Nitrophenyl-N-acetyl-b-D-galactosaminide

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₈
MDL Number: MFCD00067360
CAS Number: 14948-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR