((2-(2-(2-Bromoethoxy)ethoxy)ethoxy)methyl)benzene ≥95%

Supplier: Ambeed
Warning

A700460-5G A700460-1G
77622-496EA 298.7 USD
77622-496 77622-494
((2-(2-(2-Bromoethoxy)ethoxy)ethoxy)methyl)benzene ≥95%
((2-(2-(2-Bromoethoxy)ethoxy)ethoxy)methyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrO₃
MW: 303.1922 g/mol
MDL Number: MFCD25977587
CAS Number: 121537-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR