3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-amine ≥97%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG6-NH2

A700435-100MG A700435-1G A700435-250MG
77598-720EA 54.33 USD
77598-720 77622-490 77622-492
3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-amine ≥97%
3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-amine

Precaución: for Research Use Only

Formula: C₁₅H₂₉NO₆
MDL Number: MFCD29042376
CAS Number: 1198080-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR