pyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

Supplier: Ambeed
Danger

Synonyms: 3-Cyanopyrazolo[1,5-a]pyrimidine, Pyrazolo[1,5-a]pyrimidine-3-carbonitrile

A700079-1G A700079-250MG
77882-557EA 31 USD
77882-557 77882-558
pyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
pyrazolo[1,5-a]pyrimidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₄
MDL Number: MFCD01927799
CAS Number: 25939-87-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR