N-Butyl 3-boronobenzenesulphonamide ≥95%

Supplier: Ambeed
Warning

Synonyms: (3-(N-Butylsulphamoyl)phenyl)boronic acid

A699919-1G A699919-250MG A699919-5G
77873-100EA 79 USD
77873-100 77873-101 77873-102
N-Butyl 3-boronobenzenesulphonamide ≥95%
N-Butyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₄S
MDL Number: MFCD07783858
CAS Number: 871329-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR