Tetraammineplatinum(II) chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Tetraammineplatinum(II) chloride

A699909-100MG A699909-250MG
77669-888EA 19 USD
77669-888 77669-890
Tetraammineplatinum(II) chloride ≥98%
Tetraammineplatinum(II) chloride

Precaución: for Research Use Only

Formula: H₁₂Cl₂N₄Pt
MW: 334.11 g/mol
MDL Number: MFCD00011475
CAS Number: 13933-32-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR