4-Bromo-1-(4-fluorophenyl)-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

A699878-25G A699878-5G
77884-603EA 396 USD
77884-603 77884-604
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole ≥98%
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂
MDL Number: MFCD09801028
CAS Number: 957062-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR