(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-tert-Butyl 3-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)pyrrolidine-1-carboxylate
A699551-1G
A699551-250MG
77893-406EA
517
USD
77893-406
77893-407
(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester 95+%
(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₅H₁₈ClF₃N₂O₃ MW: 366.763 g/mol |
MDL Number:
MFCD22631739 CAS Number: 1417789-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...