1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl [3-(piperazin-1-yl)prop-1-yl]carbamate, 1-{3-[(tert-Butoxycarbonyl)amino]prop-1-yl}piperazine, tert-Butyl (3-(piperazin-1-yl)propyl)carbamate

A699532-250MG A699532-100MG A699532-1G A699532-5G
77595-084EA 26.65 USD
77595-084 77614-164 77614-166 77684-611
1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected ≥98%
1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected

Precaución: For research use only.

Formula: C₁₂H₂₅N₃O₂
MW: 243.346 g/mol
MDL Number: MFCD06808577
CAS Number: 874831-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR