1,2-Dibromonaphthalene ≥98%

Supplier: Ambeed
Warning

A699482-1G A699482-100MG A699482-250MG
77573-256EA 79 USD
77573-256 77873-098 77873-099
1,2-Dibromonaphthalene ≥98%
1,2-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
MDL Number: MFCD18072828
CAS Number: 5438-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR