9-(Ethoxyimino)-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile ≥97%

Supplier: Ambeed
Warning

Synonyms: DUB-IN-2

A699104-5MG A699104-10MG A699104-2MG
77747-405EA 22 USD
77747-405 77747-403 77747-404
9-(Ethoxyimino)-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile ≥97%
9-(Ethoxyimino)-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₅H₉N₅O
MW: 275.26 g/mol
MDL Number: MFCD28167759
CAS Number: 924296-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR