2,5,8,11,14,17-Hexaoxanonadecan-19-oic acid ≥95%

Supplier: Ambeed
Warning

A698966-1G A698966-250MG A698966-100MG A698966-5G
77598-716EA 188.6 USD
77598-716 77598-718 77622-486 77622-488
2,5,8,11,14,17-Hexaoxanonadecan-19-oic acid ≥95%
2,5,8,11,14,17-Hexaoxanonadecan-19-oic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₈
MDL Number: MFCD30536160
CAS Number: 16142-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR