MPI-5a ≥97%

Supplier: Ambeed
Danger

Synonyms: N-Hydroxy-2-(1-methyl-1H-pyrrole-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxamide, MPI_5a

A698765-10MG A698765-1MG A698765-5MG
77617-886EA 52.53 USD
77617-886 77617-888 77617-890
MPI-5a ≥97%
MPI-5a

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₃
MW: 299.32 g/mol
MDL Number: MFCD29770783
CAS Number: 1259296-46-2
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR