Silydianin ≥95%

Supplier: Ambeed
Warning

Synonyms: (3R,3aR,6R,7aR,8R)-7a-Hydroxy-8-(4-hydroxy-3-methoxyphenyl)-4-((2R,3R)-3,5,7-trihydroxy-4-oxochroman-2-yl)-3,3a,6,7a-tetrahydro-3,6-methanobenzofuran-7(2H)-one

A698317-5MG A698317-100MG A698317-10MG A698317-1MG A698317-50MG
77669-884EA 92 USD
77669-884 77873-094 77873-095 77873-096 77873-097
Silydianin ≥95%
Silydianin

Precaución: for Research Use Only

Formula: C₂₅H₂₂O₁₀
MDL Number: MFCD09970974
CAS Number: 29782-68-1
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR