2-(3-Hydroxypropyl)benzimidazole ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Benzimidazolyl)propanol, 3-(1H-Benzo[d]imidazol-2-yl)propan-1-ol, 2-Benzimidazolepropanol

A698249-1G A698249-250MG
77627-412EA 25.75 USD
77627-412 77627-414
2-(3-Hydroxypropyl)benzimidazole ≥95%
2-(3-Hydroxypropyl)benzimidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
MDL Number: MFCD00022688
CAS Number: 2403-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR