CAY10566 ≥98%

Supplier: Ambeed
Warning

A698046-10MG A698046-1MG A698046-5MG
77614-152EA 214.25 USD
77614-152 77614-154 77614-156
CAY10566 ≥98%
CAY10566

Precaución: for Research Use Only

Formula: C₁₈H₁₇ClFN₅O₂
MDL Number: MFCD11976899
CAS Number: 944808-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR