tert-Butyl (9-hydroxynonyl)carbamate ≥95%

Supplier: Ambeed
Warning

A697879-1G A697879-250MG A697879-100MG A697879-10G
77622-484EA 216.3 USD
77622-484 77783-674 77783-672 77783-673
tert-Butyl (9-hydroxynonyl)carbamate ≥95%
tert-Butyl (9-hydroxynonyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₉NO₃
MDL Number: MFCD02094529
CAS Number: 1397043-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR