Benzyl (2-(2-aminoethoxy)ethyl)carbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: N-Z-2-(2-Amino-ethoxy)-ethylamine

A697731-1G A697731-5G
77622-480EA 177.16 USD
77622-480 77622-482
Benzyl (2-(2-aminoethoxy)ethyl)carbamate ≥95%
Benzyl (2-(2-aminoethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₃
MW: 238.28 g/mol
MDL Number: MFCD18839144
CAS Number: 1290627-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR