13-Bromo-1-phenyl-2,5,8,11-tetraoxatridecane ≥95%

Supplier: Ambeed
Warning

A697573-250MG A697573-5G A697573-1G
77749-436EA 15 USD
77749-436 77749-437 77749-435
13-Bromo-1-phenyl-2,5,8,11-tetraoxatridecane ≥95%
13-Bromo-1-phenyl-2,5,8,11-tetraoxatridecane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BrO₄
MW: 347.2447 g/mol
MDL Number: MFCD31656943
CAS Number: 1239369-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR