20-Hydroxy-3,6,9,12,15,18-hexaoxaicosanoic acid ≥95%

Supplier: Ambeed
Warning

A697569-100MG A697569-250MG
77783-670EA 155 USD
77783-670 77783-671
20-Hydroxy-3,6,9,12,15,18-hexaoxaicosanoic acid ≥95%
20-Hydroxy-3,6,9,12,15,18-hexaoxaicosanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₉
MDL Number: MFCD31656911
CAS Number: 120394-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR