(2R,4S)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A696718-100MG A696718-250MG A696718-5G
77673-898EA 189 USD
77673-898 77673-900 77673-902
(2R,4S)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%
(2R,4S)-tert-Butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD11617835
CAS Number: 77450-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR