1-Bromo-2-fluoro-4-methoxy-3-nitrobenzene ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A696231-5G A696231-100MG A696231-1G A696231-250MG
77684-602EA 315 USD
77684-602 77893-396 77893-397 77893-398
1-Bromo-2-fluoro-4-methoxy-3-nitrobenzene ≥98%, Ambeed.Inc
1-Bromo-2-fluoro-4-methoxy-3-nitrobenzene

Precaución: For research use only.

Formula: C₇H₅BrFNO₃
MDL Number: MFCD20491421
CAS Number: 1352317-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR