4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 98

Supplier: Ambeed
Warning

Synonyms: 18-Nitro-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine

A695611-5G A695611-1G A695611-250MG
77669-878EA 264 USD
77669-878 77873-091 77873-092
4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 98
4'-Nitrobenzo-18-crown-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₈
MW: 357.35 g/mol
MDL Number: MFCD00143207
CAS Number: 53408-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR