2-Methoxy-4-nitrobenzaldehyde ≥98%

Supplier: Ambeed
Warning

A695405-1G A695405-250MG
77617-878EA 35.02 USD
77617-878 77747-401
2-Methoxy-4-nitrobenzaldehyde ≥98%
2-Methoxy-4-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD02683052
CAS Number: 136507-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR