2-Chloro-3,5-dimethylphenol ≥95%

Supplier: Ambeed
Warning

A695355-1G A695355-250MG
77602-764EA 154.78 USD
77602-764 77627-406
2-Chloro-3,5-dimethylphenol ≥95%
2-Chloro-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.60 g/mol
MDL Number: MFCD09835163
CAS Number: 5538-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR