2,5,8,11,14,17,20-Heptaoxadocosan-22-oic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
m-PEG6-O-CH2COOH
A695127-100MG
A695127-1G
A695127-250MG
A695127-5G
77622-472EA
17.51
USD
77622-472
77622-474
77622-476
77783-669
2,5,8,11,14,17,20-Heptaoxadocosan-22-oic acid ≥95%
2,5,8,11,14,17,20-Heptaoxadocosan-22-oic acid
Precaución: for Research Use Only
Formula:
C₁₅H₃₀O₉ |
MDL Number:
MFCD30536159 CAS Number: 75427-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...