4-Methylumbelliferyl-β-D-xylopyranoside ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-7-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

A693881-100MG A693881-250MG A693881-1G
77617-874EA 86.52 USD
77617-874 77749-425 77749-424
4-Methylumbelliferyl-β-D-xylopyranoside ≥98%
4-Methylumbelliferyl-beta-D-xylopyranoside

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₇
MW: 308.286 g/mol
MDL Number: MFCD00037607
CAS Number: 6734-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR