1,1,3,3-Tetramethyl-2-(2-oxopyridin-1(2H)-yl)isouronium hexafluorophosphate(V) ≥98%

Supplier: Ambeed
Danger

A693388-5G A693388-25G
77573-244EA 28 USD
77573-244 77886-620
1,1,3,3-Tetramethyl-2-(2-oxopyridin-1(2H)-yl)isouronium hexafluorophosphate(V) ≥98%
1,1,3,3-Tetramethyl-2-(2-oxopyridin-1(2H)-yl)isouronium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₀H₁₆F₆N₃O₂P
MDL Number: MFCD07366926
CAS Number: 364047-51-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR